Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205286.1 | NP_001192215.1 | 2 - 3 | 249 | 63 | |
NM_001205287.1 | NP_001192216.1 | 2 - 3 | 501 | 63 | ||
NM_026407.3 | NP_080683.2 | 2 - 3 | 501 | 63 | ||
GenBank mRNA | AK011646.1 | - | 2 - 3 | 475 | 63 | |
AK017817.1 | - | 2 - 3 | 459 | 63 | ||
AK133638.1 | - | 2 - 3 | 472 | 63 | ||
AK160576.1 | - | 2 - 3 | 250 | 63 | ||
AK169127.1 | - | 2 - 3 | 503 | 63 | ||
BC020318.2 | - | 2 - 3 | 456 | 5250532 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |