Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142681.1 | NP_001136153.1 | 1 - 2 | 83 | 62 | |
NM_026522.5 | NP_080798.2 | 1 - 2 | 83 | 62 | ||
GenBank mRNA | AK014071.1 | - | 1 - 2 | 32 | 62 | |
AK014141.1 | - | 1 - 2 | 58 | 62 | ||
AK036717.1 | - | 1 - 2 | 27 | 62 | ||
AK040002.1 | - | 1 - 2 | 60 | 62 | ||
AK050655.1 | - | 1 - 2 | 31 | 62 | ||
AK147377.1 | - | 1 - 2 | 46 | 62 | ||
AK160156.1 | - | 1 - 2 | 43 | 62 | ||
AK160200.1 | - | 1 - 2 | 43 | 62 | ||
BC006876.1 | - | 1 - 2 | 32 | 2647540 | 62 | |
BC061063.1 | - | 1 - 2 | 43 | 6591338 | 62 | |
BY101128.1 | - | 1 - 2 | 84 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |