Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142681.1 | NP_001136153.1 | 6 - 7 | 670 | 68 | |
NM_026522.5 | NP_080798.2 | 6 - 7 | 670 | 68 | ||
GenBank mRNA | AK014071.1 | - | 6 - 7 | 619 | 68 | |
AK036717.1 | - | 6 - 7 | 614 | 68 | ||
AK050655.1 | - | 6 - 7 | 618 | 68 | ||
AK160156.1 | - | 6 - 7 | 630 | 68 | ||
AK160200.1 | - | 6 - 7 | 630 | 68 | ||
BC006876.1 | - | 6 - 7 | 619 | 2647540 | 68 | |
BC061063.1 | - | 6 - 7 | 630 | 6591338 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |