Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2415387 |
AK019775.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2415387 |
BC145806.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2415387 |
AK015586.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2415387 |
NM_026668.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2415387 |
AK015024.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
410156 |
NM_008036.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
410156 |
X14897.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
227797 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
575611 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403431 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506580 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911252 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
637258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911985 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403324 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
92026 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
784530 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
432270 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65740 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11929 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P106986 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P104466 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2415387 |
AK019775.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2415387 |
BC145806.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2415387 |
AK015586.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2415387 |
NM_026668.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2415387 |
AK015024.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029487 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029447 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029497 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029476 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029502 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029504 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029510 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029488 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029482 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029506 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029490 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029500 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029467 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029445 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029449 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029496 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029474 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029491 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029495 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029466 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029460 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029454 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029480 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029452 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029479 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029439 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029451 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029509 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029493 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029443 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029507 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029499 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029468 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029514 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029483 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029486 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029457 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029475 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029505 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029438 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029517 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029462 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029464 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029471 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029472 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029469 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22029456 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04861610 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04861641 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P106986 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00609754 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P104466 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P452968 |
AK019775.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452968 |
NM_026668.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P452968 |
AK015024.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P102749 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125126 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P104466 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P106986 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02890577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029549 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029602 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22029474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03150531 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029441 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02762670 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02892612 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02971399 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029451 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029609 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029606 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029607 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02762671 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029456 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02892620 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02617479 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03150536 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029452 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029455 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029613 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029460 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029603 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029446 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029433 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029605 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029449 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029615 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029462 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029602 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029614 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22029611 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02910151 |
- |
- |