Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB067773.1 | - | 1 - 2 | 112 | 69 | |
AK005883.1 | - | 1 - 2 | 134 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |