Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028803.4 | NP_083079.1 | 1 - 2 | 345 | 56 | |
GenBank mRNA | AK009815.1 | - | 1 - 2 | 161 | 56 | |
AK013167.1 | - | 1 - 2 | 154 | 56 | ||
AK050365.1 | - | 1 - 2 | 232 | 56 | ||
AK050423.1 | - | 1 - 2 | 231 | 56 | ||
BC017541.1 | - | 1 - 2 | 206 | 4526961 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |