Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029269.2 | NP_083545.1 | 2 - 3 | 327 | 109 | |
GenBank mRNA | AJ315513.1 | - | 2 - 3 | 288 | 109 | |
AK002814.1 | - | 2 - 3 | 328 | 109 | ||
BC027494.1 | - | 2 - 3 | 253 | 1433655 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |