Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23189071 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23189073 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23189070 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2045268 |
BC014732.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2045268 |
BC018379.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2045268 |
BC045145.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2045268 |
BC058273.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2045268 |
AX775905.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P121774 |
BC014732.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P121774 |
BC018379.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P121774 |
BC045145.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P121774 |
BC058273.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P121774 |
AX775905.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
552259 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
973710 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
547437 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825393 |
AK122453.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825393 |
AK161919.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825393 |
BC057065.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825393 |
BC079679.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825393 |
D86214.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825393 |
NM_001042617.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825393 |
NM_012061.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
814959 |
BC119252.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
814959 |
BC119254.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
814959 |
NM_146272.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
937070 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
115472 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
204855 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
358348 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598038 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
150012 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
513996 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
733297 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
156498 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
46301 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P164386 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P148717 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2045268 |
BC014732.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2045268 |
BC018379.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2045268 |
BC045145.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2045268 |
BC058273.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2045268 |
AX775905.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P121774 |
BC014732.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P121774 |
BC018379.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P121774 |
BC045145.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P121774 |
BC058273.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P121774 |
AX775905.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189082 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189100 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189083 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189084 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189091 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189099 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189071 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189093 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189076 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189080 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189094 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189081 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189095 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189069 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189073 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189070 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189078 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189097 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189087 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23189089 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00954265 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P164386 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05350637 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P148717 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P212501 |
BC014732.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P212501 |
BC018379.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P212501 |
BC045145.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P212501 |
BC058273.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P212501 |
AK183280.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P433172 |
AF116911.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P612557 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121774 |
BC014732.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121774 |
BC018379.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121774 |
BC045145.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121774 |
BC058273.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121774 |
AX775905.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P164386 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P148717 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03746052 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03746039 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03746038 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23189070 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23189071 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23189073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189163 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189258 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189228 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189240 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189165 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189266 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189256 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189263 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23189249 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189184 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189131 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03746039 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189099 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189111 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03474350 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189095 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189116 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189107 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189093 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03553199 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189073 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03279249 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189080 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189084 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189198 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189104 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189071 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189088 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189195 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03963751 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03411447 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189190 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189121 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189118 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189082 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189076 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189134 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189186 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189135 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189138 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189120 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189081 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189114 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03889216 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189089 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189137 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189141 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189100 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189180 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189132 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189068 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03474351 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03404030 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189130 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03848305 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189109 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189189 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23189085 |
- |
- |