Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030685.3 | NP_109610.1 | 1 - 2 | 478 | 56 | |
GenBank mRNA | AB041655.1 | - | 1 - 2 | 347 | 56 | |
AK002653.1 | - | 1 - 2 | 350 | 56 | ||
AK032680.1 | - | 1 - 2 | 348 | 56 | ||
AK050508.1 | - | 1 - 2 | 318 | 56 | ||
AK075776.1 | - | 1 - 2 | 344 | 56 | ||
AK144732.1 | - | 1 - 2 | 343 | 56 | ||
AK148485.1 | - | 1 - 2 | 349 | 56 | ||
AK149786.1 | - | 1 - 2 | 348 | 56 | ||
AK150005.1 | - | 1 - 2 | 348 | 56 | ||
AK150025.1 | - | 1 - 2 | 348 | 56 | ||
AK150368.1 | - | 1 - 2 | 347 | 56 | ||
AK150428.1 | - | 1 - 2 | 348 | 56 | ||
AK150659.1 | - | 1 - 2 | 343 | 56 | ||
AK150714.1 | - | 1 - 2 | 347 | 56 | ||
AK150809.1 | - | 1 - 2 | 317 | 56 | ||
AK151323.1 | - | 1 - 2 | 317 | 56 | ||
AK151384.1 | - | 1 - 2 | 330 | 56 | ||
AK151456.1 | - | 1 - 2 | 348 | 56 | ||
AK151669.1 | - | 1 - 2 | 343 | 56 | ||
AK152373.1 | - | 1 - 2 | 343 | 56 | ||
AK152383.1 | - | 1 - 2 | 347 | 56 | ||
AK152583.1 | - | 1 - 2 | 343 | 56 | ||
AK152636.1 | - | 1 - 2 | 348 | 56 | ||
AK152806.1 | - | 1 - 2 | 348 | 56 | ||
AK152882.1 | - | 1 - 2 | 343 | 56 | ||
AK152996.1 | - | 1 - 2 | 348 | 56 | ||
AK153056.1 | - | 1 - 2 | 317 | 56 | ||
AK153118.1 | - | 1 - 2 | 317 | 56 | ||
AK153138.1 | - | 1 - 2 | 343 | 56 | ||
AK153286.1 | - | 1 - 2 | 317 | 56 | ||
AK153415.1 | - | 1 - 2 | 347 | 56 | ||
AK159271.1 | - | 1 - 2 | 348 | 56 | ||
AK161595.1 | - | 1 - 2 | 348 | 56 | ||
AK166004.1 | - | 1 - 2 | 343 | 56 | ||
AK166052.1 | - | 1 - 2 | 323 | 56 | ||
AK167482.1 | - | 1 - 2 | 318 | 56 | ||
AK167952.1 | - | 1 - 2 | 348 | 56 | ||
AK168046.1 | - | 1 - 2 | 348 | 56 | ||
AK168569.1 | - | 1 - 2 | 347 | 56 | ||
AK168734.1 | - | 1 - 2 | 347 | 56 | ||
AK168754.1 | - | 1 - 2 | 347 | 56 | ||
AK168947.1 | - | 1 - 2 | 347 | 56 | ||
AK169049.1 | - | 1 - 2 | 343 | 56 | ||
AK169175.1 | - | 1 - 2 | 348 | 56 | ||
AK170734.1 | - | 1 - 2 | 348 | 56 | ||
BC002114.1 | - | 1 - 2 | 324 | 3489738 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |