Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400006075.mm.1 |
NM_001037938.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006075.mm.1 |
NM_001037938.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006075.mm.1 |
NM_030686.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006075.mm.1 |
NM_030686.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006075.mm.1 |
TC1400000777.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006076.mm.1 |
NM_001037938.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006076.mm.1 |
NM_001037938.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006076.mm.1 |
NM_030686.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006076.mm.1 |
NM_030686.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006076.mm.1 |
TC1400000777.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006074.mm.1 |
NM_001037938.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006074.mm.1 |
NM_001037938.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006074.mm.1 |
NM_030686.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006074.mm.1 |
NM_030686.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006074.mm.1 |
TC1400000777.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006077.mm.1 |
NM_001037938.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006077.mm.1 |
NM_001037938.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006077.mm.1 |
NM_030686.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006077.mm.1 |
NM_030686.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006077.mm.1 |
TC1400000777.mm.1 |
MTA TC |