Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030719.3 | NP_109644.2 | 6 - 7 | 1037 | 66 | |
GenBank mRNA | AJ296173.1 | - | 6 - 7 | 1077 | 66 | |
AK039122.1 | - | 6 - 7 | 1063 | 66 | ||
AK052217.1 | - | 6 - 7 | 1022 | 66 | ||
BC094613.1 | - | 6 - 7 | 1037 | 6856325 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |