Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030730.2 | NP_109655.2 | 4 - 5 | 765 | 64 | |
GenBank mRNA | AJ132389.1 | - | 4 - 5 | 666 | 64 | |
AK083903.1 | - | 4 - 5 | 746 | 64 | ||
AK143506.1 | - | 3 - 4 | 381 | 64 | ||
AK155151.1 | - | 4 - 5 | 722 | 64 | ||
BC133714.1 | - | 4 - 5 | 614 | 40143565 | 64 | |
X97574.1 | - | 2 - 3 | 269 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |