Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031184.3 | NP_112461.2 | 4 - 5 | 795 | 70 | |
GenBank mRNA | AF249340.1 | - | 3 - 4 | 577 | 70 | |
AF325913.1 | - | 3 - 4 | 547 | 70 | ||
BC021517.1 | - | 4 - 5 | 624 | 5352352 | 70 | |
CN703218.1 | - | 3 - 4 | 266 | 70 | ||
CX563691.1 | - | 3 - 4 | 472 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |