Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114385.1 | NP_001107857.1 | 6 - 7 | 1041 | 75 | |
NM_031258.3 | NP_112548.2 | 6 - 7 | 1041 | 75 | ||
GenBank mRNA | AF296451.1 | - | 6 - 7 | 963 | 75 | |
AF305714.1 | - | 5 - 6 | 580 | 75 | ||
AF321853.1 | - | 5 - 6 | 517 | 75 | ||
AK142805.1 | - | 6 - 7 | 665 | 75 | ||
AK164661.1 | - | 6 - 7 | 636 | 75 | ||
BC050818.1 | - | 5 - 6 | 531 | 75 | ||
BC066832.1 | - | 6 - 7 | 675 | 30102312 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |