Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199293.1 | NP_001186222.1 | 24 - 25 | 3946 | 79 | |
NM_031386.2 | NP_113563.2 | 24 - 25 | 3927 | 79 | ||
GenBank mRNA | AK139808.1 | - | 11 - 12 | 1683 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |