Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031843.2 | NP_114031.2 | 7 - 8 | 911 | 71 | |
GenBank mRNA | AF285235.1 | - | 7 - 8 | 906 | 71 | |
AK028293.1 | - | 5 - 6 | 1532 | 71 | ||
AK140406.1 | - | 6 - 7 | 1025 | 71 | ||
AK140420.1 | - | 6 - 7 | 1025 | 71 | ||
AK154193.1 | - | 7 - 8 | 921 | 71 | ||
AK211757.1 | - | 2 - 3 | 216 | 71 | ||
BC027205.1 | - | 7 - 8 | 911 | 4190473 | 71 | |
BC052861.1 | - | 5 - 6 | 1539 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |