Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033074.3 | NP_149065.2 | 12 - 13 | 1527 | 78 | |
GenBank mRNA | AK010256.1 | - | 12 - 13 | 1494 | 78 | |
AK011146.2 | - | 12 - 13 | 1528 | 78 | ||
AK031064.1 | - | 12 - 13 | 1491 | 78 | ||
AK152945.1 | - | 12 - 13 | 1488 | 78 | ||
AK153314.1 | - | 12 - 13 | 1499 | 78 | ||
AK166693.1 | - | 12 - 13 | 1494 | 78 | ||
AK167597.1 | - | 12 - 13 | 1536 | 78 | ||
AK168969.1 | - | 12 - 13 | 1487 | 78 | ||
BC004621.1 | - | 5 - 6 | 614 | 78 | ||
BC055371.1 | - | 12 - 13 | 1513 | 30060749 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |