Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277256.1 | NP_001264185.1 | 3 - 4 | 1029 | 71 | |
NM_033524.3 | NP_277059.1 | 3 - 4 | 1029 | 71 | ||
GenBank mRNA | AB063495.1 | - | 3 - 4 | 375 | 71 | |
AK155065.1 | - | 3 - 4 | 992 | 71 | ||
AK155070.1 | - | 3 - 4 | 1031 | 71 | ||
BC041685.1 | - | 2 - 3 | 343 | 71 | ||
BC057874.1 | - | 3 - 4 | 462 | 5319749 | 71 | |
BC079606.1 | - | 3 - 4 | 650 | 30355455 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |