Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242378.1 | NP_001229307.1 | 20 - 21 | 2920 | 125 | |
NM_001242379.1 | NP_001229308.1 | 19 - 20 | 2830 | 125 | ||
NM_001242380.1 | NP_001229309.1 | 20 - 21 | 2920 | 125 | ||
NM_001242381.1 | NP_001229310.1 | 19 - 20 | 2827 | 125 | ||
NM_001242382.1 | NP_001229311.1 | 20 - 21 | 2917 | 125 | ||
NM_001242383.1 | NP_001229312.1 | 19 - 20 | 2827 | 125 | ||
NM_033552.3 | NP_291030.2 | 19 - 20 | 2830 | 125 | ||
GenBank mRNA | AB033759.1 | - | 19 - 20 | 2751 | 125 | |
AK043017.1 | - | 19 - 20 | 2806 | 125 | ||
AK220501.1 | - | 20 - 21 | 2885 | 125 | ||
BC039226.1 | - | 19 - 20 | 2779 | 4480841 | 125 | |
HM149278.1 | - | 19 - 20 | 2791 | 125 | ||
HQ533147.1 | - | 20 - 21 | 2857 | 125 | ||
HQ825316.1 | - | 20 - 21 | 2860 | 125 | ||
HQ831517.1 | - | 19 - 20 | 2767 | 125 | ||
JF500487.1 | - | 19 - 20 | 2907 | 125 | ||
JF500488.1 | - | 20 - 21 | 2997 | 125 | ||
JF500489.1 | - | 19 - 20 | 2907 | 125 | ||
JF500490.1 | - | 20 - 21 | 2997 | 125 | ||
JX220977.1 | - | 20 - 21 | 2884 | 125 | ||
JX254917.1 | - | 19 - 20 | 2797 | 125 | ||
JX268544.1 | - | 20 - 21 | 2998 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |