Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053072.3 | NP_444302.4 | 10 - 11 | 3394 | 72 | |
GenBank mRNA | AK016940.1 | - | 4 - 5 | 367 | 72 | |
AK044341.1 | - | 10 - 11 | 3392 | 72 | ||
AK173157.1 | - | 10 - 11 | 3234 | 72 | ||
BC026860.1 | - | 9 - 10 | 1732 | 72 | ||
BC139019.1 | - | 10 - 11 | 3188 | 8862041 | 72 | |
BC139022.1 | - | 10 - 11 | 3188 | 8862044 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |