Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199272.1 | NP_001186201.1 | 5 - 6 | 1045 | 66 | |
NM_053187.3 | NP_444417.2 | 4 - 5 | 1021 | 66 | ||
GenBank mRNA | AB052838.1 | - | 4 - 5 | 801 | 66 | |
AF287893.1 | - | 4 - 5 | 937 | 66 | ||
AK030427.1 | - | 5 - 6 | 641 | 66 | ||
AK030637.1 | - | 5 - 6 | 854 | 66 | ||
AK032613.1 | - | 5 - 6 | 851 | 66 | ||
BC051171.1 | - | 4 - 5 | 972 | 6307523 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |