Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053261.2 | NP_444491.1 | 3 - 4 | 525 | 101 | |
GenBank mRNA | AF353730.1 | - | 3 - 4 | 524 | 101 | |
AK154489.1 | - | 3 - 4 | 563 | 101 | ||
AK154845.1 | - | 3 - 4 | 496 | 101 | ||
AK170962.1 | - | 3 - 4 | 563 | 101 | ||
AK171626.1 | - | 3 - 4 | 498 | 101 | ||
BC011093.1 | - | 3 - 4 | 4012163 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |