Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054054.2 | NP_473395.2 | 11 - 12 | 2049 | 58 | |
GenBank mRNA | AF358660.1 | - | 11 - 12 | 2084 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |