Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054070.3 | NP_473411.2 | 6 - 7 | 652 | 74 | |
NR_028260.1 | 6 - 7 | 652 | 74 | |||
GenBank mRNA | AF329695.1 | - | 6 - 7 | 593 | 74 | |
AK159647.1 | - | 6 - 7 | 616 | 74 | ||
AK167964.1 | - | 6 - 7 | 627 | 74 | ||
AK168244.1 | - | 6 - 7 | 626 | 74 | ||
AK218541.1 | - | 2 - 3 | 122 | 74 | ||
BC056978.1 | - | 6 - 7 | 611 | 6821746 | 74 | |
BC060064.1 | - | 6 - 7 | 611 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |