Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057172.3 | NP_476513.2 | 14 - 15 | 1464 | 85 | |
GenBank mRNA | AK028418.1 | - | 14 - 15 | 1466 | 85 | |
AK032517.1 | - | 14 - 15 | 1431 | 85 | ||
AK138011.1 | - | 14 - 15 | 1409 | 85 | ||
AK160815.1 | - | 14 - 15 | 1402 | 85 | ||
AK166066.1 | - | 14 - 15 | 1409 | 85 | ||
AK167730.1 | - | 14 - 15 | 1444 | 85 | ||
AK198062.1 | - | 2 - 3 | 227 | 85 | ||
AK198700.1 | - | 2 - 3 | 227 | 85 | ||
BC014763.1 | - | 14 - 15 | 1427 | 4511225 | 85 | |
BC023780.1 | - | 14 - 15 | 1445 | 5347067 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |