Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080433.3 | NP_536681.2 | 1 - 2 | 281 | 119 | |
GenBank mRNA | AK019996.1 | - | 1 - 2 | 245 | 119 | |
BU703203.1 | - | 1 - 2 | 375 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Inventoried Probe spans exons |