Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080435.1 | NP_536683.1 | 17 - 18 | 2148 | 82 | |
GenBank mRNA | AF442771.1 | - | 17 - 18 | 2148 | 82 | |
AK141767.1 | - | 17 - 18 | 2219 | 82 | ||
AK161880.1 | - | 17 - 18 | 2167 | 82 | ||
BC015299.1 | - | 17 - 18 | 2148 | 4236007 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |