Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080440.3 | NP_536688.2 | 3 - 4 | 2491 | 62 | |
GenBank mRNA | AF453257.1 | - | 3 - 4 | 2492 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |