Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080459.2 | NP_536707.2 | 21 - 22 | 4400 | 68 | |
GenBank mRNA | AF375593.1 | - | 21 - 22 | 4400 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |