Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080575.2 | NP_542142.1 | 12 - 13 | 1789 | 114 | |
GenBank mRNA | AB046742.1 | - | 12 - 13 | 1778 | 114 | |
AK088244.1 | - | 12 - 13 | 1768 | 114 | ||
AK146568.1 | - | 12 - 13 | 1790 | 114 | ||
AK158119.1 | - | 12 - 13 | 1784 | 114 | ||
AK172412.1 | - | 12 - 13 | 1775 | 114 | ||
AK172470.1 | - | 12 - 13 | 1764 | 114 | ||
AK173275.1 | - | 12 - 13 | 1750 | 114 | ||
BC030930.1 | - | 8 - 9 | 963 | 4911679 | 114 | |
BC092278.1 | - | 12 - 13 | 1818 | 3710505 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |