Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007488.3 | NP_031514.3 | 14 - 15 | 1665 | 81 | |
GenBank mRNA | AB093225.1 | - | 16 - 17 | 1845 | 81 | |
AK032628.1 | - | 14 - 15 | 1646 | 81 | ||
BC054546.1 | - | 14 - 15 | 1586 | 6409636 | 81 | |
D63644.1 | - | 14 - 15 | 1541 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |