Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007529.2 | NP_031555.2 | 8 - 9 | 2147 | 97 | |
GenBank mRNA | AK144405.1 | - | 8 - 9 | 1948 | 97 | |
AK163646.1 | - | 1 - 2 | 208 | 97 | ||
BC052032.1 | - | 8 - 9 | 1942 | 5695027 | 97 | |
X87096.1 | - | 8 - 9 | 2023 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |