Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042527.2 | NP_001035992.1 | 14 - 15 | 2926 | 70 | |
NM_007550.4 | NP_031576.4 | 14 - 15 | 3197 | 70 | ||
GenBank mRNA | AB008674.1 | - | 14 - 15 | 3187 | 70 | |
AK137022.1 | - | 14 - 15 | 2909 | 70 | ||
AK157898.1 | - | 14 - 15 | 2926 | 70 | ||
Z98263.1 | - | 14 - 15 | 2904 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |