Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252220.1 | NP_001239149.1 | 21 - 22 | 3304 | 57 | |
NM_001252222.1 | NP_001239151.1 | 20 - 21 | 3016 | 57 | ||
NM_001252223.1 | NP_001239152.1 | 21 - 22 | 3127 | 57 | ||
NM_008676.3 | NP_032702.1 | 21 - 22 | 3127 | 57 | ||
GenBank mRNA | AF227188.1 | - | 21 - 22 | 3256 | 57 | |
AF408436.1 | - | 11 - 12 | 1851 | 57 | ||
AK077479.1 | - | 12 - 13 | 1804 | 57 | ||
AK128992.1 | - | 20 - 21 | 2808 | 57 | ||
AK129043.1 | - | 21 - 22 | 2897 | 57 | ||
AK154081.1 | - | 21 - 22 | 2913 | 57 | ||
BC020410.1 | - | 2 - 3 | 219 | 57 | ||
BC114592.1 | - | 2 - 3 | 492 | 57 | ||
BC129867.1 | - | 20 - 21 | 2779 | 40110124 | 57 | |
BC129868.1 | - | 20 - 21 | 2779 | 40110128 | 57 | |
U73039.1 | - | 21 - 22 | 2923 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |