Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008682.2 | NP_032708.2 | 12 - 13 | 1757 | 75 | |
GenBank mRNA | AK010591.1 | - | 12 - 13 | 1745 | 75 | |
AK048393.1 | - | 12 - 13 | 1709 | 75 | ||
AK153746.1 | - | 12 - 13 | 1759 | 75 | ||
BC052430.1 | - | 12 - 13 | 1726 | 6836583 | 75 | |
BC066870.1 | - | 12 - 13 | 1694 | 30476383 | 75 | |
D10712.1 | - | 12 - 13 | 1699 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |