Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163662.1 | NP_001157134.1 | 3 - 4 | 335 | 91 | |
NM_008749.2 | NP_032775.1 | 3 - 4 | 335 | 91 | ||
GenBank mRNA | AK002630.1 | - | 3 - 4 | 316 | 91 | |
AK004886.1 | - | 3 - 4 | 306 | 91 | ||
AK007976.1 | - | 3 - 4 | 309 | 91 | ||
AK045087.1 | - | 3 - 4 | 335 | 91 | ||
AK140710.1 | - | 3 - 4 | 313 | 91 | ||
AK160981.1 | - | 3 - 4 | 322 | 91 | ||
AK164088.1 | - | 3 - 4 | 317 | 91 | ||
AK170717.1 | - | 3 - 4 | 330 | 91 | ||
BC072554.1 | - | 3 - 4 | 313 | 5684979 | 91 | |
M96823.1 | - | 2 - 3 | 257 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |