Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281934.1 | NP_001268863.1 | 14 - 15 | 2654 | 67 | |
NM_008794.2 | NP_032820.2 | 14 - 15 | 2390 | 67 | ||
GenBank mRNA | AK148416.1 | - | 14 - 15 | 1975 | 67 | |
AK151980.1 | - | 14 - 15 | 1977 | 67 | ||
AK165389.1 | - | 14 - 15 | 1996 | 67 | ||
BC006730.1 | - | 13 - 14 | 1891 | 3962441 | 67 | |
U48830.1 | - | 14 - 15 | 1962 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |