Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou1f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008849.4 | NP_032875.1 | 1 - 2 | 143 | 58 | |
GenBank mRNA | AK133615.1 | - | 1 - 2 | 261 | 58 | |
BC061213.1 | - | 260 | 30252457 | 58 | ||
D12885.1 | - | 261 | 58 | |||
X57512.1 | - | 1 - 2 | 143 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |