Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008850.1 | NP_032876.1 | 3 - 4 | 425 | 65 | |
GenBank mRNA | AK144618.1 | - | 3 - 4 | 438 | 65 | |
AK149745.1 | - | 3 - 4 | 415 | 65 | ||
AK151496.1 | - | 3 - 4 | 414 | 65 | ||
AK152874.1 | - | 3 - 4 | 431 | 65 | ||
AK153569.1 | - | 3 - 4 | 438 | 65 | ||
AK165659.1 | - | 3 - 4 | 430 | 65 | ||
AK167760.1 | - | 3 - 4 | 438 | 65 | ||
AK168187.1 | - | 3 - 4 | 395 | 65 | ||
AK168720.1 | - | 3 - 4 | 428 | 65 | ||
AK187087.1 | - | 3 - 4 | 238 | 65 | ||
BC056171.1 | - | 3 - 4 | 380 | 5710926 | 65 | |
S72681.1 | - | 3 - 4 | 200 | 65 | ||
U96725.1 | - | 3 - 4 | 426 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |