Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008886.2 | NP_032912.2 | 1 - 2 | 74 | 59 | |
GenBank mRNA | AK017847.1 | - | 1 - 2 | 50 | 59 | |
AK034866.1 | - | 1 - 2 | 50 | 59 | ||
AK146365.1 | - | 1 - 2 | 87 | 59 | ||
BC066110.1 | - | 1 - 2 | 74 | 59 | ||
BC083502.1 | - | 1 - 2 | 34 | 59 | ||
BC086637.1 | - | 1 - 2 | 50 | 59 | ||
BC141287.1 | - | 1 - 2 | 50 | 9055899 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |