Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008960.2 | NP_032986.1 | 2 - 3 | 1022 | 126 | |
GenBank mRNA | AK076980.1 | - | 2 - 3 | 980 | 126 | |
AK088717.1 | - | 2 - 3 | 1001 | 126 | ||
AK164302.1 | - | 2 - 3 | 1005 | 126 | ||
BC021445.1 | - | 2 - 3 | 371 | 5038272 | 126 | |
U92437.1 | - | 2 - 3 | 1102 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |