Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008960.2 | NP_032986.1 | 6 - 7 | 1499 | 90 | |
GenBank mRNA | AK030750.1 | - | 2 - 3 | 262 | 90 | |
AK076980.1 | - | 6 - 7 | 1457 | 90 | ||
AK088717.1 | - | 6 - 7 | 1478 | 90 | ||
AK148736.1 | - | 6 - 7 | 1334 | 90 | ||
BC021445.1 | - | 6 - 7 | 848 | 5038272 | 90 | |
U92437.1 | - | 6 - 7 | 1579 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Mouse Chemokine Panel Mouse Hypoxia Panel Mouse Tumor Metastasis Panel Mouse Stem Cell Pluripotency Inventoried Mouse_Stem_Cell Probe spans exons |