Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291890.1 | NP_001278819.1 | 2 - 3 | 131 | 77 | |
NM_001291891.1 | NP_001278820.1 | 2 - 3 | 316 | 77 | ||
NM_009616.4 | NP_033746.1 | 2 - 3 | 316 | 77 | ||
GenBank mRNA | AF019887.2 | - | 2 - 3 | 221 | 77 | |
AK147217.1 | - | 2 - 3 | 239 | 77 | ||
AK147549.1 | - | 2 - 3 | 132 | 77 | ||
BC093480.1 | - | 2 - 3 | 307 | 77 | ||
CJ111483.1 | - | 2 - 3 | 316 | 77 | ||
CJ114671.1 | - | 2 - 3 | 316 | 77 | ||
D50410.2 | - | 2 - 3 | 248 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |