Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017250.1 | - | 2 - 3 | 183 | 65 | |
BC087927.1 | - | 2 - 3 | 183 | 30504437 | 65 | |
U65021.1 | - | 2 - 3 | 163 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |