Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110794.1 | NP_001104264.1 | 3 - 4 | 499 | 102 | |
NM_009674.3 | NP_033804.2 | 3 - 4 | 614 | 102 | ||
GenBank mRNA | AK032013.1 | - | 3 - 4 | 390 | 102 | |
AK077579.1 | - | 3 - 4 | 307 | 102 | ||
AK134589.1 | - | 3 - 4 | 318 | 102 | ||
AK149654.1 | - | 3 - 4 | 306 | 102 | ||
AK151438.1 | - | 3 - 4 | 308 | 102 | ||
AK161613.1 | - | 3 - 4 | 308 | 102 | ||
AK167588.1 | - | 3 - 4 | 282 | 102 | ||
AK169199.1 | - | 3 - 4 | 281 | 102 | ||
BC008997.1 | - | 3 - 4 | 298 | 3157926 | 102 | |
L13129.1 | - | 3 - 4 | 301 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |