Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037758.2 | NP_001032847.2 | 11 - 12 | 1566 | 58 | |
NM_001286465.1 | NP_001273394.1 | 11 - 12 | 1566 | 58 | ||
NM_001286466.1 | NP_001273395.1 | 10 - 11 | 1334 | 58 | ||
NM_009771.3 | NP_033901.1 | 10 - 11 | 1458 | 58 | ||
GenBank mRNA | AF081887.1 | - | 10 - 11 | 1443 | 58 | |
AF110396.1 | - | 10 - 11 | 1353 | 58 | ||
AF112979.1 | - | 10 - 11 | 1353 | 58 | ||
AK032221.1 | - | 11 - 12 | 1529 | 58 | ||
AK052317.1 | - | 12 - 13 | 1590 | 58 | ||
AK083257.1 | - | 11 - 12 | 1543 | 58 | ||
AK145091.1 | - | 10 - 11 | 1270 | 58 | ||
AK145624.1 | - | 11 - 12 | 1549 | 58 | ||
AK156660.1 | - | 10 - 11 | 1486 | 58 | ||
AK220183.1 | - | 11 - 12 | 1564 | 58 | ||
BC003989.1 | - | 10 - 11 | 1359 | 3491843 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |