Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010925.2 | NP_035055.2 | 5 - 6 | 515 | 106 | |
GenBank mRNA | AF312394.1 | - | 4 - 5 | 278 | 106 | |
AK077540.1 | - | 5 - 6 | 440 | 106 | ||
BC022910.1 | - | 4 - 5 | 381 | 4165783 | 106 | |
BC065993.1 | - | 5 - 6 | 477 | 5684672 | 106 | |
BC065994.1 | - | 5 - 6 | 435 | 5685269 | 106 | |
U79773.1 | - | 5 - 6 | 467 | 106 | ||
U79774.1 | - | 5 - 6 | 467 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |