Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111078.1 | NP_001104548.1 | 8 - 9 | 1309 | 78 | |
NM_001111079.1 | NP_001104549.1 | 8 - 9 | 1429 | 78 | ||
NM_001111080.1 | NP_001104550.1 | 8 - 9 | 1285 | 78 | ||
NM_010931.3 | NP_035061.3 | 8 - 9 | 1453 | 78 | ||
GenBank mRNA | AF274046.1 | - | 8 - 9 | 1366 | 78 | |
AK143688.1 | - | 8 - 9 | 1278 | 78 | ||
AK145376.1 | - | 8 - 9 | 1275 | 78 | ||
AK145419.1 | - | 8 - 9 | 1275 | 78 | ||
AK145543.1 | - | 8 - 9 | 1393 | 78 | ||
AK145583.1 | - | 8 - 9 | 1275 | 78 | ||
AK146951.1 | - | 8 - 9 | 1426 | 78 | ||
AK147046.1 | - | 8 - 9 | 1369 | 78 | ||
AK150489.1 | - | 8 - 9 | 1285 | 78 | ||
AK151701.1 | - | 8 - 9 | 1275 | 78 | ||
AK151837.1 | - | 8 - 9 | 1285 | 78 | ||
AK152930.1 | - | 8 - 9 | 1285 | 78 | ||
AK153083.1 | - | 8 - 9 | 1285 | 78 | ||
D87908.1 | - | 8 - 9 | 1364 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |