Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111078.1 | NP_001104548.1 | 11 - 12 | 1626 | 79 | |
NM_001111079.1 | NP_001104549.1 | 11 - 12 | 1746 | 79 | ||
NM_001111080.1 | NP_001104550.1 | 11 - 12 | 1602 | 79 | ||
NM_010931.3 | NP_035061.3 | 11 - 12 | 1770 | 79 | ||
GenBank mRNA | AF274046.1 | - | 11 - 12 | 1683 | 79 | |
AK075819.1 | - | 1 - 2 | 86 | 79 | ||
AK143688.1 | - | 11 - 12 | 1595 | 79 | ||
AK145376.1 | - | 11 - 12 | 1592 | 79 | ||
AK145419.1 | - | 11 - 12 | 1592 | 79 | ||
AK145543.1 | - | 11 - 12 | 1710 | 79 | ||
AK145583.1 | - | 11 - 12 | 1592 | 79 | ||
AK146951.1 | - | 11 - 12 | 1743 | 79 | ||
AK147046.1 | - | 11 - 12 | 1686 | 79 | ||
AK150489.1 | - | 11 - 12 | 1602 | 79 | ||
AK151701.1 | - | 11 - 12 | 1592 | 79 | ||
AK151837.1 | - | 11 - 12 | 1602 | 79 | ||
AK152930.1 | - | 11 - 12 | 1602 | 79 | ||
AK153083.1 | - | 11 - 12 | 1602 | 79 | ||
BC022167.1 | - | 11 - 12 | 1698 | 4017286 | 79 | |
D87908.1 | - | 11 - 12 | 1681 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |