Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011060.4 | NP_035190.3 | 6 - 7 | 690 | 69 | |
GenBank mRNA | AB013849.1 | - | 6 - 7 | 691 | 69 | |
AK220522.1 | - | 5 - 6 | 932 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |